siRNA Details
| Gene Symbol: ZNF827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001306215.1 | NP_001293144.1 | 15 | 7067 | ||
| NM_178835.4 | NP_849157.2 | 15 | 7076 | |||
| XM_011531634.2 | XP_011529936.1 | 14 | 7081 | |||
| XM_011531635.2 | XP_011529937.1 | 15 | 12568 | |||
| XM_011531645.2 | XP_011529947.1 | 10 | 4819 | |||
| XM_017007768.1 | XP_016863257.1 | 14 | 7076 | |||
| XM_017007769.1 | XP_016863258.1 | 14 | 13327 | |||
| XM_017007770.1 | XP_016863259.1 | 13 | 6848 | |||
| XM_017007777.1 | XP_016863266.1 | 11 | 4818 | |||
| GenBank mRNA | BC033248.1 | - | 1 | 2230 | ||

ZNF827
Genomic Map