siRNA Details
| Gene Symbol: LOC105372751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135519.1 | 3 | 592 | |||
| GenBank mRNA | BX538159.1 | - | 3 | 552 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000415182.1 | - | 3 | 551 | Chr. 21 - 24428781 - 24547942 [+] on Build GRCh38 | |

LOC105372751
Genomic Map