siRNA Details
| Gene Symbol: LOC105372751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135519.1 | 3 | 1356 | |||
| GenBank mRNA | BX538159.1 | - | 3 | 1316 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105372751| Gene Symbol: LOC105372751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135519.1 | 3 | 1356 | |||
| GenBank mRNA | BX538159.1 | - | 3 | 1316 | ||