siRNA Details
| Gene Symbol: ZNF814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC026064.2 | - | 1 | 293 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE796049.1 | - | 1 | 273 | Chr. 19 - 57885923 - 57889493 [-] on Build GRCh38 | |

ZNF814
Genomic Map