siRNA Details
| Gene Symbol: LOC388406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126009.1 | 2, 3 | 182 | |||
| GenBank mRNA | AK131012.1 | - | 2, 3 | 183 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000592917.1 | - | 2, 3 | 181 | Chr. 17 - 60564547 - 60586623 [+] on Build GRCh38 | |

LOC388406
Genomic Map