siRNA Details
| Gene Symbol: LOC388406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126009.1 | 4 | 399 | |||
| GenBank mRNA | AK131012.1 | - | 4 | 400 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC388406| Gene Symbol: LOC388406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126009.1 | 4 | 399 | |||
| GenBank mRNA | AK131012.1 | - | 4 | 400 | ||