siRNA Details
Gene Symbol: LOC105377146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135532.1 | 1 | 41 | |||
GenBank mRNA | AK131014.1 | - | 1 | 31 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU852719.1 | - | 1 | 44 | Chr. 3 - 68500637 - 68505875 [+] on Build GRCh38 |
