siRNA Details
| Gene Symbol: LOC105377146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135532.1 | 2 | 577 | |||
| GenBank mRNA | AK131014.1 | - | 2 | 567 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU852719.1 | - | 2 | 475 | Chr. 3 - 68500637 - 68505875 [+] on Build GRCh38 | |

LOC105377146
Genomic Map