siRNA Details
| Gene Symbol: LOC105377146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135532.1 | 2 | 365 | |||
| GenBank mRNA | AK131014.1 | - | 2 | 355 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105377146| Gene Symbol: LOC105377146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135532.1 | 2 | 365 | |||
| GenBank mRNA | AK131014.1 | - | 2 | 355 | ||