siRNA Details
| Gene Symbol: TUBGCP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011521454.2 | XP_011519756.1 | 17 | 6233 | ||
| XM_011521455.2 | XP_011519757.1 | 18 | 6248 | |||
| XM_017022078.1 | XP_016877567.1 | 17 | 6230 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TUBGCP4| Gene Symbol: TUBGCP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011521454.2 | XP_011519756.1 | 17 | 6233 | ||
| XM_011521455.2 | XP_011519757.1 | 18 | 6248 | |||
| XM_017022078.1 | XP_016877567.1 | 17 | 6230 | |||