siRNA Details
| Gene Symbol: TUBGCP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011521454.2 | XP_011519756.1 | 17 | 5630 | ||
| XM_011521455.2 | XP_011519757.1 | 18 | 5645 | |||
| XM_017022078.1 | XP_016877567.1 | 17 | 5627 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC100296.1 | - | 1 | 265 | Chr. 15 - 43408397 - 43409510 [+] on Build GRCh38 | |

TUBGCP4
Genomic Map