siRNA Details
| Gene Symbol: HIVEP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038260.1 | 3 | 779 | |||
| NR_038261.1 | 4 | 597 | ||||
| GenBank mRNA | BC040942.1 | - | 3 | 623 | ||
| BC046120.1 | - | 4 | 597 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIVEP3| Gene Symbol: HIVEP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038260.1 | 3 | 779 | |||
| NR_038261.1 | 4 | 597 | ||||
| GenBank mRNA | BC040942.1 | - | 3 | 623 | ||
| BC046120.1 | - | 4 | 597 | |||