siRNA Details
| Gene Symbol: LOC100131289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038929.1 | 3 | 607 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK130456.1 | - | 2 | 1212 | Chr. 6 - 27761503 - 27763267 [+] on Build GRCh38 | |

LOC100131289
Genomic Map