siRNA Details
| Gene Symbol: LOC101927884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110281.1 | 3 | 822 | |||
| GenBank mRNA | BC018659.1 | - | 3 | 826 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927884| Gene Symbol: LOC101927884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110281.1 | 3 | 822 | |||
| GenBank mRNA | BC018659.1 | - | 3 | 826 | ||