siRNA Details
| Gene Symbol: MEAF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270875.1 | NP_001257804.1 | 7 | 4643 | ||
| NM_001270876.1 | NP_001257805.1 | 6 | 4316 | |||
| NM_022756.5 | NP_073593.2 | 8 | 4673 | |||
| NR_073090.1 | 9 | 4731 | ||||
| NR_073091.1 | 8 | 4701 | ||||
| NR_073092.1 | 9 | 4762 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF172817.1 | - | 1 | 22 | Chr. 1 - 37487118 - 37490126 [-] on Build GRCh38 | |

MEAF6
Genomic Map