siRNA Details
| Gene Symbol: LOC100505564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055529.1 | - | 2 | 1044 | ||
| Gene Symbol: LOC100289656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BM929699.1 | - | 1 | 74 | ||

LOC100505564
Genomic Map