siRNA Details
| Gene Symbol: LOC338694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104161.1 | 3 | 813 | |||
| GenBank mRNA | BC043531.1 | - | 3 | 813 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC338694| Gene Symbol: LOC338694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104161.1 | 3 | 813 | |||
| GenBank mRNA | BC043531.1 | - | 3 | 813 | ||