siRNA Details
| Gene Symbol: C11orf95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB231750.1 | - | 2 | 269 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BP425126.1 | - | 1 | 53 | Chr. 11 - 63763428 - 63763943 [-] on Build GRCh38 | |

C11orf95
Genomic Map