siRNA Details
| Gene Symbol: LOC101928565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF087964.1 | - | 1 | 99 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928565| Gene Symbol: LOC101928565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF087964.1 | - | 1 | 99 | ||