siRNA Details
| Gene Symbol: FUNDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF267862.1 | - | 1 | 52 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA258633.1 | - | 1 | 52 | Chr. 5 - 73508509 - 73508886 [+] on Build GRCh38 | |

FUNDC2
Genomic Map