siRNA Details
| Gene Symbol: LYST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102436.2 | 4 | 2219 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LYST| Gene Symbol: LYST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102436.2 | 4 | 2219 | |||