siRNA Details
Gene Symbol: GHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242404.2 | NP_001229333.1 | 1 | 67 | ||
GenBank mRNA | Z11849.1 | - | 1 | 67 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242404.2 | NP_001229333.1 | 1 | 67 | ||
GenBank mRNA | Z11849.1 | - | 1 | 67 |