siRNA Details
| Gene Symbol: LOC101927780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110416.1 | 4 | 1566 | |||
| GenBank mRNA | BX648502.1 | - | 4 | 1568 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927780| Gene Symbol: LOC101927780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110416.1 | 4 | 1566 | |||
| GenBank mRNA | BX648502.1 | - | 4 | 1568 | ||