siRNA Details
| Gene Symbol: LOC101928251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135635.1 | 2 | 2232 | |||
| GenBank mRNA | BC041474.1 | - | 2 | 2230 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928251| Gene Symbol: LOC101928251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135635.1 | 2 | 2232 | |||
| GenBank mRNA | BC041474.1 | - | 2 | 2230 | ||