siRNA Details
Gene Symbol: LOC101928251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135635.1 | 2 | 325 | |||
GenBank mRNA | BC041474.1 | - | 2 | 323 | ||
BI919218.1 | - | 2 | 355 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000583547.2 | - | 2 | 396 | Chr. 17 - 73513685 - 73518770 [+] on Build GRCh38 |
