siRNA Details
| Gene Symbol: CLSTN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017007022.1 | XP_016862511.1 | 17 | 9440 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK056386.1 | - | 1 | 889 | Chr. 3 - 140571706 - 140575229 [+] on Build GRCh38 | |

CLSTN2
Genomic Map