siRNA Details
| Gene Symbol: TTC28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011530022.1 | XP_011528324.1 | 14 | 4574 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AL122113.1 | - | 1 | 2621 | Chr. 22 - 28020309 - 28023353 [-] on Build GRCh38 | |

TTC28
Genomic Map