siRNA Details
Gene Symbol: TTC28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011530019.2 | XP_011528321.1 | 15 | 4627 | ||
XM_011530021.2 | XP_011528323.1 | 15 | 4684 | |||
XM_011530022.1 | XP_011528324.1 | 14 | 4959 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL122113.1 | - | 1 | 3006 | Chr. 22 - 28020309 - 28023353 [-] on Build GRCh38 |
