siRNA Details
Gene Symbol: CAST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U31346.1 | - | 12 | 1155 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK026822.1 | - | 1 | 1779 | Chr. 5 - 96777650 - 96779982 [+] on Build GRCh38 |
