siRNA Details
| Gene Symbol: CMIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017023733.1 | XP_016879222.1 | 1 | 1811 | ||
| GenBank mRNA | AK127550.1 | - | 1 | 1811 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CMIP| Gene Symbol: CMIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017023733.1 | XP_016879222.1 | 1 | 1811 | ||
| GenBank mRNA | AK127550.1 | - | 1 | 1811 | ||