siRNA Details
| Gene Symbol: RBM26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK027772.1 | - | 1 | 556 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK021576.1 | - | 2 | 1610 | Chr. 13 - 79316229 - 79350237 [-] on Build GRCh38 | |

RBM26
Genomic Map