siRNA Details
Gene Symbol: GCLM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308253.1 | NP_001295182.1 | 6 | 3831 | ||
NM_002061.3 | NP_002052.1 | 7 | 3897 | |||
XM_011541261.2 | XP_011539563.1 | 7 | 3698 | |||
XM_017001056.1 | XP_016856545.1 | 8 | 4099 | |||
XM_017001057.1 | XP_016856546.1 | 7 | 4033 | |||
GenBank mRNA | BF513627.1 | - | 1 | 472 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR598364.1 | - | 1 | 27 | Chr. 1 - 93885236 - 93886379 [-] on Build GRCh38 |
