siRNA Details
| Gene Symbol: TRAPPC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011510354.2 | XP_011508656.1 | 7 | 2393 | ||
| GenBank mRNA | AK094194.1 | - | 1 | 2170 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRAPPC12| Gene Symbol: TRAPPC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011510354.2 | XP_011508656.1 | 7 | 2393 | ||
| GenBank mRNA | AK094194.1 | - | 1 | 2170 | ||