siRNA Details
| Gene Symbol: TRAPPC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011510354.2 | XP_011508656.1 | 7 | 3031 | ||
| GenBank mRNA | AK094194.1 | - | 1 | 2808 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM717927.1 | - | 1 | 295 | Chr. 2 - 3453531 - 3453934 [+] on Build GRCh38 | |

TRAPPC12
Genomic Map