siRNA Details
| Gene Symbol: LOC105376360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131187.1 | 2 | 2173 | |||
| GenBank mRNA | BC037918.1 | - | 3 | 1984 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105376360| Gene Symbol: LOC105376360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131187.1 | 2 | 2173 | |||
| GenBank mRNA | BC037918.1 | - | 3 | 1984 | ||