siRNA Details
| Gene Symbol: LOC284939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK094442.1 | - | 1 | 891 | ||
| Gene Symbol: MAPK8IP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011530679.2 | XP_011528981.1 | 12 | 4174 | ||
| XM_011530680.2 | XP_011528982.1 | 12 | 4060 | |||
| XM_011530681.2 | XP_011528983.1 | 12 | 4039 | |||

LOC284939
Genomic Map