siRNA Details
| Gene Symbol: LOC101927494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135563.1 | 2 | 776 | |||
| GenBank mRNA | BC035252.1 | - | 2 | 776 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927494| Gene Symbol: LOC101927494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135563.1 | 2 | 776 | |||
| GenBank mRNA | BC035252.1 | - | 2 | 776 | ||