siRNA Details
| Gene Symbol: EBF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022659.3 | NP_073150.2 | 16 | 4464 | ||
| GenBank mRNA | AK093878.1 | - | 1 | 1789 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EBF2| Gene Symbol: EBF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022659.3 | NP_073150.2 | 16 | 4464 | ||
| GenBank mRNA | AK093878.1 | - | 1 | 1789 | ||