siRNA Details
| Gene Symbol: ZNF326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320185.1 | NP_001307114.1 | 12 | 4460 | ||
| NM_181781.3 | NP_861446.2 | 10 | 4263 | |||
| NM_182976.3 | NP_892021.1 | 12 | 4727 | |||
| XM_005270779.4 | XP_005270836.1 | 11 | 4301 | |||
| XM_011541290.2 | XP_011539592.1 | 12 | 4433 | |||
| GenBank mRNA | AK124539.1 | - | 1 | 756 | ||

ZNF326
Genomic Map