siRNA Details
Gene Symbol: SNU13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028811.1 | XP_016884300.1 | 3 | 968 | ||
GenBank mRNA | AK124020.1 | - | 1 | 1211 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SNU13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028811.1 | XP_016884300.1 | 3 | 968 | ||
GenBank mRNA | AK124020.1 | - | 1 | 1211 |