siRNA Details
| Gene Symbol: SNU13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017028811.1 | XP_016884300.1 | 3 | 636 | ||
| GenBank mRNA | AK124020.1 | - | 1 | 879 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG822290.1 | - | 1 | 401 | Chr. 22 - 41675038 - 41688328 [-] on Build GRCh38 | |

SNU13
Genomic Map