siRNA Details
| Gene Symbol: ADIPOQ-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046662.1 | 1 | 266 | |||
| GenBank mRNA | BC036509.1 | - | 2 | 400 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADIPOQ-AS1| Gene Symbol: ADIPOQ-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046662.1 | 1 | 266 | |||
| GenBank mRNA | BC036509.1 | - | 2 | 400 | ||