siRNA Details
Gene Symbol: ADIPOQ-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046662.1 | 1 | 1241 | |||
GenBank mRNA | BC036509.1 | - | 2 | 1377 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ADIPOQ-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046662.1 | 1 | 1241 | |||
GenBank mRNA | BC036509.1 | - | 2 | 1377 |