siRNA Details
| Gene Symbol: COMMD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092477.1 | - | 1 | 521 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA843501.1 | - | 1 | 521 | Chr. 8 - 144839360 - 144839944 [-] on Build GRCh38 | |

COMMD5
Genomic Map