siRNA Details
| Gene Symbol: LOC101928160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134306.1 | 6 | 1234 | |||
| GenBank mRNA | BC035363.1 | - | 6 | 1234 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928160| Gene Symbol: LOC101928160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134306.1 | 6 | 1234 | |||
| GenBank mRNA | BC035363.1 | - | 6 | 1234 | ||