siRNA Details
| Gene Symbol: LOC101928160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134306.1 | 6 | 603 | |||
| GenBank mRNA | BC035363.1 | - | 6 | 603 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI829323.1 | - | 6 | 601 | Chr. 8 - 143708051 - 143713266 [+] on Build GRCh38 | |

LOC101928160
Genomic Map