siRNA Details
Gene Symbol: LOC101928160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134306.1 | 6 | 1000 | |||
GenBank mRNA | BC035363.1 | - | 6 | 1000 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134306.1 | 6 | 1000 | |||
GenBank mRNA | BC035363.1 | - | 6 | 1000 |