siRNA Details
| Gene Symbol: LOC101927694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104648.1 | 2 | 921 | |||
| GenBank mRNA | AK125770.1 | - | 2 | 921 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000541258.1 | - | 2 | 921 | Chr. 12 - 128118217 - 128121952 [+] on Build GRCh38 | |

LOC101927694
Genomic Map