siRNA Details
| Gene Symbol: UBE2I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC014997.1 | - | 1 | 1168 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBE2I| Gene Symbol: UBE2I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC014997.1 | - | 1 | 1168 | ||