siRNA Details
| Gene Symbol: TTLL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006712875.2 | XP_006712938.1 | 20 | 6025 | ||
| XM_017005387.1 | XP_016860876.1 | 20 | 6203 | |||
| XM_017005388.1 | XP_016860877.1 | 21 | 6090 | |||
| XM_017005389.1 | XP_016860878.1 | 21 | 6109 | |||
| XM_017005390.1 | XP_016860879.1 | 20 | 6043 | |||
| XM_017005391.1 | XP_016860880.1 | 19 | 6108 | |||
| XM_017005392.1 | XP_016860881.1 | 18 | 6277 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK126797.1 | - | 1 | 568 | Chr. 2 - 218755964 - 218759387 [+] on Build GRCh38 | |

TTLL4
Genomic Map